siRNA Details
Gene Symbol: Aplp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012906.1 | NP_037038.1 | 9, 10 | 1570 | ||
NM_022520.1 | NP_071965.1 | 9, 10 | 1570 | |||
GenBank mRNA | FM030557.1 | - | 4, 5 | 436 | ||
X77934.1 | - | 9, 10 | 1558 |
