siRNA Details
Gene Symbol: Dap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022526.2 | NP_071971.1 | 2, 3 | 351 | ||
GenBank mRNA | BC060569.1 | - | 2, 3 | 169 | 6890457 | |
FQ218203.1 | - | 2, 3 | 236 | |||
FQ228698.1 | - | 2, 3 | 189 | |||
FQ228865.1 | - | 2, 3 | 195 | |||
U05334.1 | - | 2, 3 | 352 |
