siRNA Details
Gene Symbol: Dap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022526.2 | NP_071971.1 | 4 | 440 | ||
GenBank mRNA | BC060569.1 | - | 4 | 258 | 6890457 | |
FQ218203.1 | - | 4 | 325 | |||
FQ228698.1 | - | 4 | 278 | |||
FQ228865.1 | - | 4 | 284 | |||
U05334.1 | - | 4 | 441 |
