siRNA Details
Gene Symbol: Sec14l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022608.1 | NP_072130.1 | 10, 11 | 941 | ||
GenBank mRNA | AJ132352.1 | - | 10, 11 | 956 | ||
BC162038.1 | - | 10, 11 | 941 | 7099625 |
