siRNA Details
Gene Symbol: Ntsr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022695.2 | NP_073186.1 | 4 | 1230 | ||
XM_006239918.2 | XP_006239980.1 | 4 | 1017 | |||
GenBank mRNA | X97121.1 | - | 4 | 1232 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ntsr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022695.2 | NP_073186.1 | 4 | 1230 | ||
XM_006239918.2 | XP_006239980.1 | 4 | 1017 | |||
GenBank mRNA | X97121.1 | - | 4 | 1232 |
Set Membership: |
![]() |