siRNA Details
Gene Symbol: Ntsr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022695.2 | NP_073186.1 | 1 | 310 | ||
XM_006239918.2 | XP_006239980.1 | 1 | 278 | |||
GenBank mRNA | AY946024.1 | - | 1 | 272 | ||
X97121.1 | - | 1 | 312 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
