siRNA Details
Gene Symbol: Flot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022701.2 | NP_073192.2 | 6 | 610 | ||
XM_006256024.2 | XP_006256086.1 | 6 | 629 | |||
GenBank mRNA | BC064652.1 | - | 6 | 609 | 5598180 | |
CK359020.1 | - | 6 | 610 | |||
DY569467.1 | - | 3 | 292 | |||
U60976.1 | - | 6 | 540 | |||
U60977.1 | - | 5 | 614 |
