siRNA Details
Gene Symbol: Agap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023026.2 | NP_075415.2 | 5 | 1406 | ||
XM_006241499.2 | XP_006241561.1 | 5 | 2157 | |||
XM_006241500.2 | XP_006241562.1 | 5 | 753 | |||
XM_008765421.1 | XP_008763643.1 | 5 | 759 | |||
GenBank mRNA | AF280816.1 | - | 5 | 1406 | ||
AY128688.1 | - | 5 | 1406 |
