siRNA Details
Gene Symbol: Gbgt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023094.3 | NP_075582.3 | 4, 5 | 231 | ||
GenBank mRNA | AB081651.1 | - | 4, 5 | 281 | ||
AF264018.2 | - | 4, 5 | 231 | |||
AF469945.1 | - | 4, 5 | 231 |
