siRNA Details
Gene Symbol: Nup62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023098.2 | NP_075586.1 | 2 | 1414 | ||
XM_006229150.2 | XP_006229212.1 | 2 | 1613 | |||
XM_006229151.2 | XP_006229213.1 | 2 | 2319 | |||
GenBank mRNA | BC128709.1 | - | 1 | 1318 | 8363131 | |
J04143.1 | - | 1 | 172 | |||
S75997.1 | - | 351 | ||||
X52583.1 | - | 1 | 1994 |
