siRNA Details
Gene Symbol: Nup62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023098.2 | NP_075586.1 | 2 | 258 | ||
XM_006229150.2 | XP_006229212.1 | 2 | 457 | |||
XM_006229151.2 | XP_006229213.1 | 2 | 1163 | |||
GenBank mRNA | BC128709.1 | - | 1 | 162 | 8363131 | |
CK600248.1 | - | 2 | 258 | |||
X52583.1 | - | 1 | 838 |
