siRNA Details
Gene Symbol: LOC654482 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039174.1 | NP_001034263.1 | 3 | 223 | ||
XM_006242497.2 | XP_006242559.1 | 3 | 691 | |||
GenBank mRNA | BC079425.1 | - | 3 | 223 | 7114230 |
