siRNA Details
Gene Symbol: Crisp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039393.2 | NP_001034482.2 | 8 | 814 | ||
XM_008766935.1 | XP_008765157.1 | 8 | 814 | |||
GenBank mRNA | DQ200907.1 | - | 8 | 814 |
