siRNA Details
Gene Symbol: Arl6ip5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023972.3 | NP_076462.1 | 2 | 216 | ||
GenBank mRNA | AF240182.1 | - | 2 | 216 | ||
BC061548.1 | - | 228 | 6922366 | |||
FQ213351.1 | - | 249 | ||||
FQ213919.1 | - | 238 | ||||
FQ216190.1 | - | 249 | ||||
FQ216551.1 | - | 250 | ||||
FQ225681.1 | - | 249 | ||||
FQ228780.1 | - | 314 | ||||
FQ232358.1 | - | 249 |
