siRNA Details
Gene Symbol: Arl6ip5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023972.3 | NP_076462.1 | 1 | 80 | ||
GenBank mRNA | AF240182.1 | - | 1 | 80 | ||
BC061548.1 | - | 92 | 6922366 | |||
FQ213351.1 | - | 113 | ||||
FQ213919.1 | - | 102 | ||||
FQ216190.1 | - | 113 | ||||
FQ216551.1 | - | 114 | ||||
FQ225681.1 | - | 113 | ||||
FQ228780.1 | - | 178 | ||||
FQ232358.1 | - | 113 |
