siRNA Details
Gene Symbol: Slc6a19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039722.2 | NP_001034811.1 | 10 | 1446 | ||
GenBank mRNA | AJ890206.1 | - | 10 | 1518 | ||
EF474455.1 | - | 10 | 1479 |

More Information
Other Species Targeted

Gene Ontology Categories:
