siRNA Details
Gene Symbol: Mapk11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109532.2 | NP_001103002.2 | 11 | 1038 | ||
XM_008765792.1 | XP_008764014.1 | 10 | 1163 | |||
GenBank mRNA | BC162032.1 | - | 12 | 1074 | 9102947 |
