siRNA Details
Gene Symbol: Slc17a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271214.1 | NP_001258143.1 | 11 | 1842 | ||
XM_008771657.1 | XP_008769879.1 | 11 | 1847 | |||
XM_008771658.1 | XP_008769880.1 | 12 | 2161 | |||
XM_008771659.1 | XP_008769881.1 | 12 | 2231 | |||
XM_008771660.1 | XP_008769882.1 | 10 | 1613 |
