siRNA Details
Gene Symbol: Aup1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079899.1 | NP_001073368.1 | 2 | 179 | ||
XM_008763029.1 | XP_008761251.1 | 2 | 264 | |||
GenBank mRNA | BC127544.1 | - | 2 | 179 | 7319831 | |
FQ228919.1 | - | 2 | 212 |
