siRNA Details
Gene Symbol: Mbnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111064.1 | NP_001104534.1 | 8, 9 | 1788 | ||
GenBank mRNA | AI602815.1 | - | 4, 5 | 323 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_207515, NM_175341, AK085709, BC079898, AK159213, and others.
Gene Ontology Categories:
