siRNA Details
Gene Symbol: Lcn12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128138.1 | NP_001121610.1 | 1 | 258 | ||
GenBank mRNA | DQ537492.1 | - | 2 | 224 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lcn12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128138.1 | NP_001121610.1 | 1 | 258 | ||
GenBank mRNA | DQ537492.1 | - | 2 | 224 |