siRNA Details
Gene Symbol: Spcs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191073.1 | NP_001178002.1 | 5 | 444 | ||
GenBank mRNA | AY724480.1 | - | 563 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Spcs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191073.1 | NP_001178002.1 | 5 | 444 | ||
GenBank mRNA | AY724480.1 | - | 563 |