siRNA Details
Gene Symbol: Mlip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766417.1 | XP_008764639.1 | 4 | 1786 | ||
XM_008766418.1 | XP_008764640.1 | 3 | 1630 | |||
XM_008766420.1 | XP_008764642.1 | 4 | 1594 | |||
GenBank mRNA | BC092625.1 | - | 1290 | |||
FQ224108.1 | - | 1449 |
