siRNA Details
Gene Symbol: Noxred1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750205.3 | XP_003750253.1 | 3, 4 | 651 | ||
XM_006240401.2 | XP_006240463.1 | 3, 4 | 467 | |||
XM_006240402.2 | XP_006240464.1 | 3, 4 | 664 | |||
XM_006240403.2 | XP_006240465.1 | 3, 4 | 797 | |||
XM_008764893.1 | XP_008763115.1 | 4, 5 | 548 | |||
XM_008764894.1 | XP_008763116.1 | 4, 5 | 762 |
