siRNA Details
Gene Symbol: Ppp2r3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139492.1 | NP_001132964.1 | 3 | 1026 | ||
GenBank mRNA | BC161890.1 | - | 3 | 1026 | 7135877 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ppp2r3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139492.1 | NP_001132964.1 | 3 | 1026 | ||
GenBank mRNA | BC161890.1 | - | 3 | 1026 | 7135877 |