siRNA Details
Gene Symbol: LOC682102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134732.1 | NP_001128204.1 | 2 | 518 | ||
GenBank mRNA | CK653557.1 | - | 2 | 556 | ||
CV103711.1 | - | 2 | 386 |

More Information
Other Species Targeted

