siRNA Details
Gene Symbol: Txlna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127633.1 | NP_001121105.1 | 11 | 1542 | ||
XM_006238972.2 | XP_006239034.1 | 11 | 1549 | |||
GenBank mRNA | BC166486.1 | - | 11 | 1542 | 7099983 |
