siRNA Details
Gene Symbol: Sept6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257463.2 | XP_006257525.1 | 1 | 280 | ||
XM_008773441.1 | XP_008771663.1 | 1 | 280 | |||
XM_008773442.1 | XP_008771664.1 | 1 | 280 | |||
XM_008773443.1 | XP_008771665.1 | 1 | 280 |
