siRNA Details
Gene Symbol: Galnt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172063.2 | NP_001165534.1 | 9, 10 | 1763 | ||
XM_006242294.2 | XP_006242356.1 | 11, 12 | 2849 | |||
XM_006242295.2 | XP_006242357.1 | 11, 12 | 2172 | |||
XM_006242296.2 | XP_006242358.1 | 11, 12 | 2116 | |||
XM_008765651.1 | XP_008763873.1 | 12, 13 | 2351 | |||
XM_008765652.1 | XP_008763874.1 | 11, 12 | 2108 | |||
XM_008765653.1 | XP_008763875.1 | 11, 12 | 2388 | |||
GenBank mRNA | FQ211776.1 | - | 11, 12 | 2140 |
