siRNA Details
Gene Symbol: Sin3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115038.1 | NP_001108510.1 | 5, 6 | 791 | ||
GenBank mRNA | BC158844.1 | - | 5, 6 | 704 | 7442664 | |
FQ220173.1 | - | 5, 6 | 714 | |||
FQ220358.1 | - | 5, 6 | 714 | |||
FQ220509.1 | - | 5, 6 | 714 | |||
FQ220717.1 | - | 5, 6 | 714 | |||
FQ229609.1 | - | 5, 6 | 714 | |||
FQ229881.1 | - | 5, 6 | 714 |
