siRNA Details
Gene Symbol: Zfp639 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080907.2 | NP_001074376.1 | 6 | 1613 | ||
XM_006232254.2 | XP_006232316.1 | 7 | 1683 | |||
XM_008760942.1 | XP_008759164.1 | 6 | 3162 | |||
XM_008760943.1 | XP_008759165.1 | 7 | 2434 | |||
XM_008760944.1 | XP_008759166.1 | 4 | 2204 | |||
GenBank mRNA | BC087704.1 | - | 6 | 1614 | 7313723 |
