siRNA Details
Gene Symbol: Lbh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129880.2 | NP_001123352.1 | 2 | 306 | ||
XM_008764533.1 | XP_008762755.1 | 4 | 455 | |||
GenBank mRNA | CB744030.1 | - | 2 | 359 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lbh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129880.2 | NP_001123352.1 | 2 | 306 | ||
XM_008764533.1 | XP_008762755.1 | 4 | 455 | |||
GenBank mRNA | CB744030.1 | - | 2 | 359 |