siRNA Details
Gene Symbol: Lbh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129880.2 | NP_001123352.1 | 3 | 477 | ||
XM_008764533.1 | XP_008762755.1 | 5 | 626 |

More Information
Other Species Targeted

Gene Ontology Categories:
