siRNA Details
Gene Symbol: Ipo11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760732.1 | XP_008758954.1 | 2 | 254 |
Gene Symbol: LOC314407 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764901.1 | XP_008763123.1 | 1 | 156 |
