siRNA Details
Gene Symbol: Nipal3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006239277.2 | XP_006239339.1 | 10 | 1253 | ||
XM_578498.5 | XP_578498.3 | 10 | 1320 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nipal3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006239277.2 | XP_006239339.1 | 10 | 1253 | ||
XM_578498.5 | XP_578498.3 | 10 | 1320 |