siRNA Details
Gene Symbol: Sik3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766173.1 | XP_008764395.1 | 10 | 1295 | ||
XM_008766174.1 | XP_008764396.1 | 10 | 1294 | |||
XM_008766175.1 | XP_008764397.1 | 10 | 1294 |
