siRNA Details
Gene Symbol: LOC102548162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228202.2 | XP_006228264.1 | 2, 3 | 1504 | ||
XM_006228203.2 | XP_006228265.1 | 2, 3 | 1502 | |||
XM_008759003.1 | XP_008757225.1 | 2, 3 | 1502 |
