siRNA Details
Gene Symbol: Rcor3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134985.1 | NP_001128457.1 | 11 | 1304 | ||
GenBank mRNA | BC169033.1 | - | 11 | 1304 | 7114893 |
Gene Symbol: LOC102551140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006250501.1 | XP_006250563.1 | 183 |
