siRNA Details
Gene Symbol: Mtif3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115041.2 | NP_001108513.2 | 3 | 536 | ||
XM_006248837.2 | XP_006248899.1 | 3 | 573 | |||
GenBank mRNA | BC158794.1 | - | 3 | 536 | 7314259 |
