siRNA Details
Gene Symbol: Cenpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750123.2 | XP_003750171.1 | 4 | 438 | ||
XM_006239855.1 | XP_006239917.1 | 4 | 438 | |||
XM_008764544.1 | XP_008762766.1 | 4 | 504 | |||
XM_008764545.1 | XP_008762767.1 | 4 | 504 | |||
XM_008764546.1 | XP_008762768.1 | 4 | 504 | |||
GenBank mRNA | FQ232123.1 | - | 2 | 401 | ||
FQ233391.1 | - | 2 | 401 |
Gene Symbol: LOC102551278 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006251433.1 | XP_006251495.1 | 1 | 398 |

More Information
Other Species Targeted

Gene Ontology Categories:
