siRNA Details
Gene Symbol: Cenpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750123.2 | XP_003750171.1 | 6 | 884 | ||
XM_008764545.1 | XP_008762767.1 | 6 | 974 | |||
XM_008764546.1 | XP_008762768.1 | 6 | 950 |
