siRNA Details
Gene Symbol: Echdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101010.1 | NP_001094480.1 | 275 | |||
GenBank mRNA | BC101897.1 | - | 275 | 7932594 | ||
FQ219501.1 | - | 313 |

More Information
Other Species Targeted

Gene Ontology Categories:
