siRNA Details
Gene Symbol: Zfpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286936.1 | NP_001273865.1 | 3 | 244 | ||
XM_006230901.2 | XP_006230963.1 | 3 | 220 | |||
XM_008760175.1 | XP_008758397.1 | 3 | 208 | |||
GenBank mRNA | CB717504.1 | - | 3 | 244 | ||
FQ227385.1 | - | 3 | 280 |
