siRNA Details
Gene Symbol: Zfpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286936.1 | NP_001273865.1 | 3, 4 | 317 | ||
XM_006230901.2 | XP_006230963.1 | 3, 4 | 293 | |||
XM_008760175.1 | XP_008758397.1 | 3, 4 | 281 | |||
GenBank mRNA | CB717504.1 | - | 3, 4 | 317 | ||
DY546174.1 | - | 1, 2 | 37 | |||
FQ227385.1 | - | 3, 4 | 353 |
