siRNA Details
Gene Symbol: Zfpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286936.1 | NP_001273865.1 | 7 | 799 | ||
XM_006230901.2 | XP_006230963.1 | 7 | 775 | |||
XM_008760175.1 | XP_008758397.1 | 7 | 763 | |||
GenBank mRNA | BU758664.1 | - | 2 | 518 | ||
DY546174.1 | - | 5 | 519 | |||
FQ227385.1 | - | 7 | 835 |
