siRNA Details
Gene Symbol: Cdca5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115042.2 | NP_001108514.2 | 6, 7 | 887 | ||
XM_008760177.1 | XP_008758399.1 | 2, 3 | 547 | |||
GenBank mRNA | BC097411.1 | - | 6, 7 | 817 | ||
BC158663.1 | - | 6, 7 | 887 | 8369770 | ||
BC158764.1 | - | 6, 7 | 826 | 7111047 |
