siRNA Details
Gene Symbol: Cabyr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143893.1 | NP_001137365.1 | 2 | 203 | ||
XM_006254426.2 | XP_006254488.1 | 2 | 2256 | |||
XM_008771955.1 | XP_008770177.1 | 2 | 2256 | |||
GenBank mRNA | BC160837.1 | - | 2 | 187 |
