siRNA Details
Gene Symbol: Il31ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257278.2 | NP_001244207.1 | 15 | 2177 | ||
XM_008760740.1 | XP_008758962.1 | 15 | 2349 | |||
GenBank mRNA | JQ013736.1 | - | 15 | 2177 |
