siRNA Details
Gene Symbol: Il3ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248989.2 | XP_006249051.1 | 4 | 2756 | ||
GenBank mRNA | BP502982.1 | - | 1 | 32 | ||
FM042837.1 | - | 4 | 463 | |||
FM046911.1 | - | 3 | 237 | |||
FQ214452.1 | - | 1 | 59 |
