siRNA Details
Gene Symbol: Il3ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248989.2 | XP_006249051.1 | 5 | 2879 | ||
GenBank mRNA | BP502982.1 | - | 1 | 289 | ||
FM046911.1 | - | 4 | 360 | |||
FQ214452.1 | - | 1 | 316 |
